D-(+)-Glucose 1 M in aqueous solution, sterile-filtered

Supplier: Thermo Scientific Chemicals

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

J60067-AK J60067-EQE
CAAAJ60067-AKEA 4523.86 CAD
CAAAJ60067-AK CAAAJ60067-EQE
D-(+)-Glucose 1 M in aqueous solution, sterile-filtered
D(+)-Glucose

Liquid


CAS No.: 50-99-7 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Liquid MDL No.: MFCD00148912

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR