HEPES, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60064-AE J60064-AK J60463-AE J60463-AK J61047-AE J61047-AK J61275-AE J61275-AK J63002-AE J63002-AK J63218-AE J63218-AK
CAAAJ60064-AEEA 4086.26 CAD
CAAAJ60064-AE CAAAJ60064-AK CAAAJ60463-AE CAAAJ60463-AK CAAAJ61047-AE CAAAJ61047-AK CAAAJ61275-AE CAAAJ61275-AK CAAAJ63002-AE CAAAJ63002-AK CAAAJ63218-AE CAAAJ63218-AK CAAAJ63614-AE CAAAJ63614-AK
HEPES, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 7365-45-9
MDL No.: MFCD00006158

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR