ADA, 0.2 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60054-AE J61176-AE J61176-AK J61259-AE J61259-AK J62424-AE J62424-AK
CAAAJ60054-AEEA 96.25 CAD
CAAAJ60054-AE CAAAJ60054-AK CAAAJ61176-AE CAAAJ61176-AK CAAAJ61259-AE CAAAJ61259-AK CAAAJ62424-AE CAAAJ62424-AK
ADA, 0.2 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

N-(2-Acetimido)iminodiacetic acid monosodium salt (ADA) buffer solutions PH values 6.0 to 7.5.


CAS No.: 7415-22-7
Appearance: Colorless
MDL No.: MFCD00065483


Caution: Storage Temperature: 2 to 8°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR