N-Lauroylsarcosine sodium salt 95%

Supplier: Thermo Scientific Chemicals
Danger

J60040-18 J60040-09
CAAAJ60040-18EA 1085.81 CAD
CAAAJ60040-18 CAAAJ60040-09
N-Lauroylsarcosine sodium salt 95%
N-Lauroylsarcosine sodium salt

Used in the cell lysis step of RNA purification. Anionic detergent


Crystalline powder


Used in the cell lysis step of RNA purification. Anionic detergent

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Melting Pt: 44…47 °C
Flash Pt: ≥ 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
EINECS: 205-281-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,4368
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR