Enrofloxacin

Supplier: Thermo Scientific Chemicals

J60023-06 J60023-14
CAAAJ60023-06EA 204.91 CAD
CAAAJ60023-06 CAAAJ60023-14
Enrofloxacin
Enrofloxacin

A plant fungicide and antibacterial agent


Powder


Applications: A plant fungicide and antibacterial agent

Formula: C₁₉H₂₂FN₃O₃
Melting Pt: 221…226 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792463
CAS Number: 93106-60-6
Merck Index: 14,3592

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR