Thiamine nitrate
Supplier: Thermo Scientific Chemicals
J60014-09
J60014-14
CAAAJ60014-09EA
109.35
CAD
CAAAJ60014-09
CAAAJ60014-14
Thiamine nitrate
Thiamine nitrate
Powder
CAS No.: 532-43-4
Molecular Formula: C12H17N5O4S
Formula Weight: 327.36
Physical Form: Powder
Appearance: White
Melting Point: 190°
MDL No.: MFCD00036330
Formula:
C₁₂H₁₇N₅O₄S Melting Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036330 CAS Number: 532-43-4 EINECS: 208-537-4 Merck Index: 14,9295 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...