Naloxone hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

J60013-03 J60013-MC J60013-ME
CAAAJ60013-03EA 1242.09 CAD
CAAAJ60013-03 CAAAJ60013-MC CAAAJ60013-ME
Naloxone hydrochloride 98%
Naloxone hydrochloride

A specific opiate antagonist. Blocks the action of sigma-agonists at opioid sites


Crystalline powder


Applications: A specific opiate antagonist. Blocks the action of sigma-agonists at opioid sites

Formula: C₁₉H₂₂ClNO₄
Melting Pt: 200…205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069322
CAS Number: 357-08-4
EINECS: 206-611-0
Merck Index: 14,6362

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR