Carbon black ≥99% (metals basis), Super P® Conductive
Supplier: Thermo Scientific Chemicals
Synonyms:
Furnace black, Lamp Black
H30253-14
H30253-22
AAH30253-14EA
468.28
CAD
AAH30253-14
AAH30253-22
Carbon black ≥99% (metals basis), Super P® Conductive
Carbon black
Purity: 99+%
CAS No.: 1333-86-4
Molecular Formula: C
Formula Weight: 12.01
Odor: Odorless
UN No.: 1325
MDL No.: MFCD00133992
Formula:
C MW: 12.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 1333-86-4 EINECS: 215-609-9 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...