Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(pentamethylcyclopentadienyl)cobalticinium hexafluorophosphate

H27761-03 H27761-06
AAH27761-03EA 161.16 CAD
AAH27761-03 AAH27761-06
Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate 98%
Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate

Purity: 98% CAS No.: 79973-42-5 Molecular Formula: C20H30CoF6P Formula Weight: 474.36 Melting Point: ca 305° dec. UN No.: 3261 MDL No.: MFCD00269818

Formula: C₂₀H₃₀CoF₆P
MW: 474.35 g/mol
Melting Pt: ca 305 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00269818
CAS Number: 79973-42-5
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR