(4-(Hydroxymethyl)phenyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27531-03
H27531-06
CAAAH27531-03EA
68.57
CAD
CAAAH27531-03
CAAAH27531-06
(4-(Hydroxymethyl)phenyl)acetic acid 97%
(4-(Hydroxymethyl)phenyl)acetic acid
Purity: 98%
CAS No.: 73401-74-8
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 131-134°
MDL No.: MFCD00065692
Formula:
C₉H₁₀O₃ Melting Pt: 131…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065692 CAS Number: 73401-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...