Titanium(IV) 2-ethylhexyloxide ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
77107-22
77107-36
77107-A3
CAAA77107-22EA
40.61
CAD
CAAA77107-22
CAAA77107-36
CAAA77107-A3
Titanium(IV) 2-ethylhexyloxide ≥95%
Titanium(IV) 2-ethylhexyloxide
Liquid
Purity: 95%
CAS No.: 1070-10-6
Molecular Formula: C32H68O4Ti
Formula Weight: 564.79
Flash Point: 71°(160°F)
Physical Form: Liquid
Density: 0.926
Refractive Index: 1.482
Sensitivity: Moisture Sensitive
Appearance: Pale yellow
Melting Point:
Formula:
C₃₂H₆₈O₄Ti Boiling Pt: 194 °C (0.25 mmHg) Melting Pt: < –25 °C Density: 0,926 Flash Pt: 71 °C (160 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015264 CAS Number: 1070-10-6 EINECS: 213-969-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...