Copper ≥99.8% (metals basis), foil, annealed, uncoated, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
046986-RG
046986-RH
046986-RF
AA46986-RGEA
162.74
CAD
AA46986-RG
AA46986-RH
AA46986-RF
Copper ≥99.8% (metals basis), foil, annealed, uncoated, Thickness: 0.025 mm (0.001 in)
Copper
ca. 20.2g/30x30cm
Purity: 99.8%
CAS No.: 7440-50-8
Physical Form: ≈20.2g/30x30cm
Appearance: Copper
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...