Palladium 10% on activated carbon paste, unreduced 10R394

Supplier: Thermo Scientific Chemicals

45558-06 45558-14
CAAA45558-06EA 259.12 CAD
CAAA45558-06 CAAA45558-14
Palladium 10% on activated carbon paste, unreduced 10R394
Palladium
Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR