Gallium(III) ethanolate
Supplier: Thermo Scientific Chemicals
|
Warning
|
44859-03
44859-06
CAAAA44859-03EA
164.32
CAD
CAAAA44859-03
CAAAA44859-06
Gallium(III) ethanolate
Gallium(III) ethanolate
Liquid
CAS No.: 2572-25-0
Molecular Formula: C6H15GaO3.xHOCH2CH3
Formula Weight: 204.90
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.25
Appearance: Yellow
MDL No.: MFCD01866730
Formula:
C₆H₁₅GaO₃ Density: 1,25 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01866730 CAS Number: 2572-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...