Aluminium oxide catalyst support low SiO₂

Supplier: Thermo Scientific Chemicals

44849-A1 44849-A7
CAAAA44849-A1EA 267.02 CAD
CAAAA44849-A1 CAAAA44849-A7
Aluminium oxide catalyst support low SiO₂
Aluminium oxide

3 mm Spheres


Physical Form: 3 mm Spheres MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR