Barium-2-ethylhexanoate (∼17.5% Ba) in 2-ethylhexanoic acid , 99.8%

Supplier: Thermo Scientific Chemicals
Danger

44847-06 44847-14
CAAAA44847-06EA 38.08 CAD
CAAAA44847-06 CAAAA44847-14 CAAAA44847-22
Barium-2-ethylhexanoate (∼17.5% Ba) in 2-ethylhexanoic acid , 99.8%
Barium 2-ethylhexanoate

Viscous liquid


CAS No.: 2457-01-4 Molecular Formula: C16H30BaO4 Formula Weight: 423.76 Physical Form: Viscous liquid Appearance: Yellow

Formula: C₁₆H₃₀BaO₄
Storage Temperature: Ambient
MDL Number: MFCD00058696
CAS Number: 2457-01-4
EINECS: 219-535-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR