Niobium(V) oxalate hydrate

Supplier: Thermo Scientific Chemicals
Warning

44819-14 44819-22 44819-36
CAAAA44819-14EA 89.43 CAD
CAAAA44819-14 CAAAA44819-22 CAAAA44819-36
Niobium(V) oxalate hydrate
Niobium(V) oxalate hydrate

Powder


CAS No.: 21348-59-4 Molecular Formula: C10H5NbO20.xH2O Formula Weight: 538.04(anhy) Physical Form: Powder Odor: Odorless MDL No.: MFCD00050743

Formula: C₁₀H₅NbO₂₀·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00050743
CAS Number: 21348-59-4
EINECS: 244-341-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR