Hexamethyldisilathiane, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
44818-03
44818-06
CAAAA44818-03EA
85.79
CAD
CAAAA44818-03
CAAAA44818-06
Hexamethyldisilathiane, tech.
Hexamethyldisilathiane
Liquid in ampoules
CAS No.: 3385-94-2
Molecular Formula: C6H18SSi2
Formula Weight: 178.44
Flash Point: 26°(79°F)
Physical Form: Liquid in ampoules
Density: 0.851
Refractive Index: 1.4598
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Pungent
Boiling Point: 162-166°
Beilstein Reference: 1740046
UN No.: 1993
MDL No.: MFCD00014851
Formula:
C₆H₁₈SSi₂ Boiling Pt: 162…166 °C Density: 0,851 Flash Pt: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014851 CAS Number: 3385-94-2 EINECS: 222-201-4 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...