5'-O-(4,4'-Dimethoxytrityl)thymidine ≥98%

Supplier: Thermo Scientific Chemicals
Warning

44815-02 44815-04
CAAAA44815-02EA 225.94 CAD
CAAAA44815-02 CAAAA44815-04 CAAAA44815-09 CAAAA44815-77
5'-O-(4,4'-Dimethoxytrityl)thymidine ≥98%
5'-O-(4,4'-Dimethoxytrityl)thymidine

Powder


Purity: 98+% CAS No.: 40615-39-2 Molecular Formula: C31H32N2O7 Formula Weight: 544.60 Physical Form: Powder MDL No.: MFCD00010113

Formula: C₃₁H₃₂N₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00010113
CAS Number: 40615-39-2
EINECS: 255-003-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR