Hydroxypropyl methylcellulose (Hypromellose), 40 - 60 mPa.s (2% in H2O)

Supplier: Thermo Scientific Chemicals

44779-18 44779-30 44779-A1
CAAAA44779-18EA 71.1 CAD
CAAAA44779-18 CAAAA44779-30 CAAAA44779-A1
Hydroxypropyl methylcellulose (Hypromellose), 40 - 60 mPa.s (2% in H2O)
Hydroxypropyl methyl cellulose (Hypromellose)

Powder


CAS No.: 9004-65-3 Physical Form: Powder Merck Reference: 14,4842 MDL No.: MFCD00131360


Caution: Incompatible with strong oxidizing agents, bleaching powder, fluorine and nitric acid.

Formula: C₃H₈O₂·xCH₄O
Storage Temperature: Ambient
MDL Number: MFCD00131360
CAS Number: 9004-65-3
Merck Index: 14,4842

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR