Poly(3-octylthiophene-2,5-diyl) regiorandom low metals

Supplier: Thermo Scientific Chemicals

44778-01 44778-03
CAAAA44778-01EA 355.5 CAD
CAAAA44778-01 CAAAA44778-03
Poly(3-octylthiophene-2,5-diyl) regiorandom low metals
Poly(3-octylthiophene-2,5-diyl)

Powder


Applications: Conducting polymers


Applications: Conducting polymers

Storage Temperature: Ambient
MDL Number: MFCD00241412
CAS Number: 104934-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR