Allyl cyanide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
44775-14
44775-22
CAAAA44775-14EA
96.06
CAD
CAAAA44775-14
CAAAA44775-22
Allyl cyanide 97%
Allyl cyanide
Liquid
Purity: 97%
CAS No.: 109-75-1
Molecular Formula: C4H5N
Formula Weight: 67.09
Flash Point: 24°(75°F)
Physical Form: Liquid
Density: 0.834
Refractive Index: 1.4050
Appearance: Colorless to pale brown
Odor: Unpleasant
Melting Point: -87°
Boiling Point: 118-120°
Beilstein Reference: 605352
Merck Reference: 14,290
UN No.: 1992
MDL No.: MFCD00001962
Formula:
C₄H₅N MW: 67.09 g/mol Boiling Pt: 118…120 °C Melting Pt: –87 °C Density: 0.834 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001962 CAS Number: 109-75-1 EINECS: 203-701-1 UN: 1992 ADR: 3 III Merck Index: 14,290 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...