D-Biotin N-Succinimidyl Ester ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide

44771-01 44771-03
CAAAA44771-01EA 322.32 CAD
CAAAA44771-01 CAAAA44771-03 CAAAA44771-MF
D-Biotin N-Succinimidyl Ester ≥98%
D-Biotin-N-succinimidyl ester

Powder


Applications: In coupling reactions to amino acids, peptides and proteins through primary amines


Applications: In coupling reactions to amino acids, peptides and proteins through primary amines

Formula: C₁₄H₁₉N₃O₅S
Melting Pt: 212…215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078531
CAS Number: 35013-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR