Zirconium(IV) Propoxide ≥70% (w/w) in n-propanol, ChemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
44770-18
44770-30
CAAAA44770-18EA
59.88
CAD
CAAAA44770-18
CAAAA44770-30
Zirconium(IV) Propoxide ≥70% (w/w) in n-propanol, ChemSeal™
Zirconium(IV) Propoxide
Packaged under Argon in resealable ChemSeal™ bottles
Liquid
CAS No.: 23519-77-9
Molecular Formula: Zr[O(CH2)2CH3]4
Formula Weight: 327.56
Flash Point: 28°(83°F)
Physical Form: Liquid
Density: 1.044
Refractive Index: 1.4510
Sensitivity: Moisture Sensitive
UN No.: 1993
MDL No.: MFCD00015307
Formula:
C₁₂H₂₈O₄Zr Density: 1.044 Flash Pt: 28 °C (83 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015307 CAS Number: 23519-77-9 EINECS: 245-711-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...