Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
Supplier: Thermo Scientific Chemicals
44764-AD
44764-AK
44764-K2
44764-K7
CAAAA44764-ADEA
50.24
CAD
CAAAA44764-AD
CAAAA44764-AK
CAAAA44764-K2
CAAAA44764-K7
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
Polydimethylsiloxane, trimethylsiloxy terminated
Liquid
CAS No.: 9016-00-6
Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
Physical Form: Liquid
Density: 0.974
Refractive Index: 1.4035
Odor: Odorless
MDL No.: MFCD00148360
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃ Density: 0.974 Storage Temperature: Ambient |
MDL Number:
MFCD00148360 CAS Number: 9016-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...