3-Trimethylsilylpropynal ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Trimethylsilyl)-2-propynal

44763-03 44763-06
CAAAA44763-03EA 149.15 CAD
CAAAA44763-03 CAAAA44763-06
3-Trimethylsilylpropynal ≥97%
3-Trimethylsilylpropynal

Liquid, Packaged under argon


Purity: 97% CAS No.: 2975-46-4 Molecular Formula: C6H10OSi Formula Weight: 126.23 Storage Temperature: 2°C to 8°C Flash Point: 29°(84°F) Physical Form: Liquid, Packaged under argon Density: 0.86 Refractive Index: 1.4435 Sensitivity: Air and Moisture Sensitive Appearance: Colorless to yellow Boiling Point: 58-60°/37.5mm UN No.: 1993 MDL No.: MFCD01103709

Formula: C₆H₁₀OSi
MW: 126.23 g/mol
Boiling Pt: 58…60 °C (37.5 mmHg)
Density: 0,86
Flash Pt: 29 °C (84 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD01103709
CAS Number: 2975-46-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR