2-Aminoadenosine ≥97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,6-Diamino-9-(beta-D-ribofuranosyl)purine, (2R,3R,4S,5R)-2-(2,6-Diamino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2,6-Diaminopurine Riboside

44748-03 44748-06 44748-14
CAAAA44748-03EA 82.16 CAD
CAAAA44748-03 CAAAA44748-06 CAAAA44748-14
2-Aminoadenosine ≥97%
2-Aminoadenosine

Powder


Purity: 97% CAS No.: 2096-10-8 Molecular Formula: C10H14N6O4 Formula Weight: 282.26 Physical Form: Powder MDL No.: MFCD00053556

Formula: C₁₀H₁₄N₆O₄
MW: 282.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053556
CAS Number: 2096-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR