Silicon(II) oxide, high surface area catalyst support

Supplier: Thermo Scientific Chemicals
Warning

44740-22 44740-36 44740-A3
CAAAA44740-22EA 70.15 CAD
CAAAA44740-22 CAAAA44740-36 CAAAA44740-A3
Silicon(II) oxide, high surface area catalyst support
Silicon (II) oxide

3.18mm (0.125in) Pellets


Applications: Typical properties: S.A. 250m2/g


Applications: Typical properties: S.A.250m2/g

Formula: SiO
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS Number: 10097-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR