Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98%

Supplier: Thermo Scientific Chemicals
Warning

44737-04 44737-09
CAAAA44737-04EA 113.13 CAD
CAAAA44737-04 CAAAA44737-09
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98%
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid

Powder


Purity: 98% CAS No.: 869-52-3 Molecular Formula: C18H30N4O12 Formula Weight: 494.45 Physical Form: Powder MDL No.: MFCD00004290

Formula: C₁₈H₃₀N₄O₁₂
Storage Temperature: Ambient
MDL Number: MFCD00004290
CAS Number: 869-52-3
EINECS: 212-790-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR