Poly(styrene-divinylbenzene), cross-linked with 2% DVB 200-400 mesh

Supplier: Thermo Scientific Chemicals

44708-14 44708-22 44708-36
CAAAA44708-14EA 99.86 CAD
CAAAA44708-14 CAAAA44708-22 CAAAA44708-36
Poly(styrene-divinylbenzene), cross-linked with 2% DVB 200-400 mesh
Poly(styrene-divinylbenzene)

Powder
Preparation of Merrifield type resins


CAS No.: 9003-70-7 Physical Form: Powder Odor: Odorless MDL No.: MFCD00146442


Applications: Preparation of Merrifield type resins

Formula: [-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
Storage Temperature: Ambient
MDL Number: MFCD00146442
CAS Number: 9003-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR