Ethyl hydrogen malonate ≥90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid
44692-06
44692-14
44692-22
44692-36
CAAAA44692-06EA
58.14
CAD
CAAAA44692-06
CAAAA44692-14
CAAAA44692-22
CAAAA44692-36
Ethyl hydrogen malonate ≥90%
Ethyl hydrogen malonate
Liquid
Purity: 90+%
CAS No.: 1071-46-1
Molecular Formula: C5H8O4
Formula Weight: 132.12
Flash Point:
Formula:
C₅H₈O₄ MW: 132.115 g/mol Boiling Pt: 106 °C (3 mmHg) Density: 1,119 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020490 CAS Number: 1071-46-1 EINECS: 213-992-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...