Poly(acrylic acid) 25% (w/w) in water

Supplier: Thermo Scientific Chemicals

44669-22 44669-36
CAAAA44669-22EA 83.11 CAD
CAAAA44669-22 CAAAA44669-36
Poly(acrylic acid) 25% (w/w) in water
Poly(acrylic acid)

Liquid


CAS No.: 9003-01-4 Molecular Formula: [-CH2CH(CO2H)-] Physical Form: Liquid Density: 1.09 MDL No.: MFCD00084394

Formula: C₃H₄O₂
MW: 72.063 g/mol
Density: 1.09
Storage Temperature: Ambient
MDL Number: MFCD00084394
CAS Number: 9003-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR