Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000
Supplier: Thermo Scientific Chemicals
44644-18
44644-30
44644-A1
44644-A6
CAAAA44644-18EA
45.82
CAD
CAAAA44644-18
CAAAA44644-30
CAAAA44644-A1
CAAAA44644-A6
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000
Polydimethylsiloxane, trimethylsiloxy terminated
Viscous liquid
CAS No.: 9016-00-6
Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
Flash Point: 321°(610°F)
Physical Form: Viscous liquid
Density: 0.977
Refractive Index: 1.4035
Appearance: Colorless
Odor: Odorless
Melting Point:
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃ Boiling Pt: >205 °C Melting Pt: < –40 °C Density: 0.977 Flash Pt: 321 °C (610 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00148360 CAS Number: 9016-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...