Palladium, plasma standard solution, Specpure®, Pd 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
44631-AD
44631-AE
44631-AP
CAAAA44631-ADEA
172.22
CAD
CAAAA44631-AD
CAAAA44631-AE
CAAAA44631-AP
Palladium, plasma standard solution, Specpure®, Pd 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Palladium Standards
Liquid
Molecular Formula: Pd in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011167
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...