Copper(II) trifluoroacetate hydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Copper(II) 2,2,2-trifluoroacetate hydrate

44624-09 44624-18
CAAAA44624-09EA 276.5 CAD
CAAAA44624-09 CAAAA44624-18
Copper(II) trifluoroacetate hydrate
Copper(II) trifluoroacetate hydrate

Crystalline


CAS No.: 123333-88-0 Molecular Formula: Cu(OOCCF3)2.xH2O Formula Weight: 289.57(anhy) Physical Form: Crystalline MDL No.: MFCD00013200

Formula: C₄CuF₆O₄·xH₂O
MW: 289.576 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00013200
CAS Number: 123333-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR