Hydrogen chloride ∼2.5 M in ethanol

Supplier: Thermo Scientific Chemicals
Danger

044605-AK 044605-K2
AA44605-AKEA 231.53 CAD
AA44605-AK AA44605-K2
Hydrogen chloride ∼2.5 M in ethanol
Hydrogen chloride

Liquid


CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid UN No.: 2924 MDL No.: MFCD00011324

Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR