Titanium(IV) 2-ethylhexanoate ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44602-22
44602-36
CAAAA44602-22EA
445.56
CAD
CAAAA44602-22
CAAAA44602-36
Titanium(IV) 2-ethylhexanoate ≥97%
Titanium(IV) 2-ethylhexanoate
Liquid
Purity: 97%
CAS No.: 3645-34-9
Molecular Formula: C32H60O8Ti
Formula Weight: 623.75
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD05663781
Formula:
C₃₂H₆₀O₈Ti MW: 620.691 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663781 CAS Number: 3645-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...