Yttrium(III) trifluoroacetate hydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44597-09
44597-18
CAAAA44597-09EA
124.66
CAD
CAAAA44597-09
CAAAA44597-18
Yttrium(III) trifluoroacetate hydrate ≥99.9%
Yttrium(III) trifluoroacetate hydrate
Crystalline
Purity: 99.9%
CAS No.: 304851-95-4
Molecular Formula: Y(OOCCF3)3.xH2O
Formula Weight: 427.95(anhy)
Physical Form: Crystalline
MDL No.: MFCD05663780
Formula:
C₆F₉O₆Y·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD05663780 CAS Number: 304851-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...