Diammonium wolframate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Ammonium tungstate

44588-09 44588-18 44588-30
CAAAA44588-09EA 164.32 CAD
CAAAA44588-09 CAAAA44588-18 CAAAA44588-30
Diammonium wolframate ≥99.99% (metals basis)
di-Ammonium wolframate

Powder


Purity: 99.99+% CAS No.: 11140-77-5 Molecular Formula: (NH4)2WO4 Formula Weight: 283.91 Physical Form: Powder Solubility: Soluble in warm water MDL No.: MFCD00015956

Formula: H₈N₂O₄W
MW: 283.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015956
CAS Number: 11140-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR