Neodymium(III) acetate hydrate ≥99.9%

Supplier: Thermo Scientific Chemicals

Synonyms: Neodymium(III) acetate xhydrate, Neodymium triacetate hydrate

44587-18 44587-30
CAAAA44587-18EA 110.6 CAD
CAAAA44587-18 CAAAA44587-30
Neodymium(III) acetate hydrate ≥99.9%
Neodymium(III) acetate hydrate

Crystalline


Purity: 99.9% CAS No.: 334869-71-5 Molecular Formula: Nd(OOCCH3)3.xH2O Formula Weight: 321.38(anhy) Physical Form: Crystalline MDL No.: MFCD00013040

Formula: C₆H₁₁NdO₇
MW: 339.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013040
CAS Number: 334869-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR