Ruthenium 2% on alumina, pellets

Supplier: Thermo Scientific Chemicals

44575-14 44575-22 44575-36
CAAAA44575-14EA 222.78 CAD
CAAAA44575-14 CAAAA44575-22 CAAAA44575-36
Ruthenium 2% on alumina, pellets
Ruthenium

S.A. nominally 200m2/g


CAS No.: 7440-18-8 Physical Form: S.A. nominally 200m2/g Odor: Odorless MDL No.: MFCD00011207

Formula: Ru
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR