Indium(III) isopropoxide 5% (w/v) in 2-propanol , 99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44562-AC
44562-AE
CAAAA44562-ACEA
213.3
CAD
CAAAA44562-AC
CAAAA44562-AE
Indium(III) isopropoxide 5% (w/v) in 2-propanol , 99% (metals basis)
Indium (III) isopropoxide
Liquid
Purity: 99+%
CAS No.: 38218-24-5
Molecular Formula: C9H21InO3
Formula Weight: 292.08
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 0.78
Sensitivity: Moisture Sensitive
Appearance: Colorless
Boiling Point: 82°
UN No.: 1219
MDL No.: MFCD00210318
Formula:
C₉H₂₁InO₃ MW: 292.08 g/mol Boiling Pt: 82 °C Density: 0.78 Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210318 CAS Number: 38218-24-5 UN: 1219 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...