N-Phenyl-p-toluidine ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44560-09
44560-18
CAAAA44560-09EA
102.38
CAD
CAAAA44560-09
CAAAA44560-18
N-Phenyl-p-toluidine ≥98%
N-Phenyl-p-toluidine
Crystalline
Purity: 98%
CAS No.: 620-84-8
Molecular Formula: C13H13N
Formula Weight: 183.25
Flash Point: 155°(311°F)
Physical Form: Crystalline
Melting Point: 85-86°
Boiling Point: 334°
MDL No.: MFCD00092921
Formula:
C₁₃H₁₃N Boiling Pt: 334 °C Melting Pt: 85…86 °C Flash Pt: 155 °C (311 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00092921 CAS Number: 620-84-8 EINECS: 210-655-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...