1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chlorodimethyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl)silane
44543-04
44543-09
CAAAA44543-04EA
108.39
CAD
CAAAA44543-04
CAAAA44543-09
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane 97%
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Liquid
Purity: 97%
CAS No.: 102488-47-1
Molecular Formula: C10H10ClF13Si
Formula Weight: 440.71
Flash Point: 52°(180°F)
Physical Form: Liquid
Density: 1.461
Refractive Index: 1.3453
Sensitivity: Moisture Sensitive
Appearance: Colorless
Boiling Point: 189-191°
Beilstein Reference: 7040835
UN No.: 2986
MDL No.: MFCD00042359
Formula:
C₁₀H₁₀ClF₁₃Si MW: 440.71 Boiling Pt: 189…191 °C Density: 1,461 Flash Pt: 52 °C (180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042359 CAS Number: 102488-47-1 UN: 2986 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...