Retinoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin
44540-02
44540-04
44540-77
CAAAA44540-02EA
158
CAD
CAAAA44540-02
CAAAA44540-04
CAAAA44540-77
Retinoic acid 98%
Retinoic acid
Crystalline
Purity: 98%
CAS No.: 302-79-4
Molecular Formula: C20H28O2
Formula Weight: 300.44
Physical Form: Crystalline
Sensitivity: Light Sensitive
Beilstein Reference: 2057223
Merck Reference: 14,8165
MDL No.: MFCD00001551
Formula:
C₂₀H₂₈O₂ Melting Pt: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001551 CAS Number: 302-79-4 EINECS: 206-129-0 Merck Index: 14,8165 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...