Poly(styrene) atactic, M.W. 125,000-250,000
Supplier: Thermo Scientific Chemicals
44537-09
44537-18
CAAAA44537-09EA
52.77
CAD
CAAAA44537-09
CAAAA44537-18
Poly(styrene) atactic, M.W. 125,000-250,000
Poly(styrene)
Beads
CAS No.: 9003-53-6
Formula Weight: 125,000-250,000
Physical Form: Beads
Odor: Odorless
MDL No.: MFCD00084450
Formula:
C₈H₈ Storage Temperature: Ambient |
MDL Number:
MFCD00084450 CAS Number: 9003-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...