Titanium(IV) oxide 20 - 35% in water, colloidal dispersion
Supplier: Thermo Scientific Chemicals
|
Warning
|
44517-36
44517-A1
44517-A7
CAAAA44517-36EA
152.63
CAD
CAAAA44517-36
CAAAA44517-A1
CAAAA44517-A7
Titanium(IV) oxide 20 - 35% in water, colloidal dispersion
Titanium (IV) oxide
Applications: UV protection in clear coatings
CAS No.: 13463-67-7
Molecular Formula: TiO2
Formula Weight: 79.90
Density: 1.26
Boiling Point: 368°
MDL No.: MFCD00011269
Applications: UV protection in clear coatings
Formula:
TiO₂ Boiling Pt: 368 °C Density: 1.26 Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 13463-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...