Fullerene buckytube/nanotube single-walled

Supplier: Thermo Scientific Chemicals
Warning

44508-01 44508-03
CAAAA44508-01EA 110.13 CAD
CAAAA44508-01 CAAAA44508-03 CAAAA44508-06
Fullerene buckytube/nanotube single-walled
Fullerene buckytube/nanotube

Powder


CAS No.: 308068-56-6 Physical Form: Powder Appearance: Black UN No.: 1325 MDL No.: MFCD00282904

Storage Temperature: Ambient
MDL Number: MFCD00282904
CAS Number: 308068-56-6
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR