Tebbe Reagent 0.5 M in toluene
Supplier: Thermo Scientific Chemicals
|
Danger
|
44506-AA
44506-AC
CAAAA44506-AAEA
119.13
CAD
CAAAA44506-AA
CAAAA44506-AC
Tebbe Reagent 0.5 M in toluene
Tebbe Reagent
Liquid
CAS No.: 67719-69-1
Molecular Formula: C13H18AlClTi
Formula Weight: 284.62
Flash Point: 1°(34°F)
Physical Form: Liquid
Density: 0.927
Sensitivity: Air and Moisture Sensitive
Appearance: Dark red-brown
UN No.: 3399
MDL No.: MFCD00151575
Formula:
C₁₃H₁₈AlClTi Density: 0.927 Flash Pt: 1 °C (34 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00151575 CAS Number: 67719-69-1 UN: 3399 ADR: 4.3,II Merck Index: 14,9091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...