2-(Hydroxymethyl)-15-crown-5 98%

Supplier: Thermo Scientific Chemicals
Warning

44500-03 44500-06
CAAAA44500-01EA 118.45 CAD
CAAAA44500-01 CAAAA44500-03 CAAAA44500-06
2-(Hydroxymethyl)-15-crown-5 98%
2-(Hydroxymethyl)-15-crown-5

Liquid


Purity: 98% CAS No.: 75507-25-4 Molecular Formula: C11H22O6 Formula Weight: 250.29 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.175 Refractive Index: 1.479 Boiling Point: 135°/1.5mm Beilstein Reference: 3589706 MDL No.: MFCD00192215

Formula: C₁₁H₂₂O₆
Boiling Pt: 135 °C (1.5 mmHg)
Density: 1,175
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192215
CAS Number: 75507-25-4
EINECS: 278-222-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR